Ampicillin sodium salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

0219014825 0219014880
IC19014825EA 295.59 USD
IC19014825 IC19014880
Ampicillin sodium salt
Ampicillin sodium salt

Ampicillin is a semi-synthetic derivative of penicillin, active as a broad-spectrum antibiotic which is used to select for ampicillin resistance in mutated and transformed cells.


  • pKa=2.5 (-COOH), 7.3 (-NH2) at 25 °C
  • Loss on drying: <2%
  • pH: 8 - 10 (1% aqeuous solution)


Ampicillin is a β-lactam antibiotic with an amino group attached to the penicillin structure


Ampicillin is inactivated by beta-lactamases and acts against gram-positive and gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines.


It inhibits bacterial cell-wall synthesis in E. coli (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It is bactericidal only to growing Escherichia coli. It is resisted by the cleavage of β-lactam ring of ampicillin by β-lactamase.


White to Off-White powder


Soluble in water (50 mg/mL); soluble in 1 M Ammonium Hydroxide, dilute acids or bases; practically insoluble in alcohol, chloroform, ether and fixed oils

Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
Melting Pt: 215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f8c861541ff88be68be667c8edf9a10a1b866a&vid=e2f8c861541ff88be68be667c8edf9a10a1b866a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR