Ampicillin, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219014605 0219014625 0219014680
IC19014605EA 191.22 USD
IC19014605 IC19014625 IC19014680
Ampicillin, powder
Ampicillin

Ampicillin is a semi-synthetic derivative of penicillin, active as a broad-spectrum antibiotic which is used to select bacteria cells with specific resistance during general microbiology studies.


  • pKa=2.5 (-COOH); 7.3 (-NH2) at 25 °C
  • pH: 4.51


Ampicillin is a β-lactam antibiotic with an amino group attached to the penicillin structure


Ampicillin is inactivated by beta-lactamases and acts against gram-positive and gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines. This antibiotic is used to study antibiotic resistance and penetration limitations, the synergy between multiple antibiotics, certain bloodstream infections, and has been used to develop PCR assays to detect resistance genes in cerebrospinal fluid.


It inhibits bacterial cell-wall synthesis in E. coli (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It is bactericidal only to growing Escherichia coli. It is resisted by the cleavage of β-lactam ring of ampicillin by β-lactamase.


White powder


Soluble in 1 M Ammonium Hydroxide (50 mg/mL), dilute acids or bases, slightly soluble in water, practically insoluble in alcohol, chloroform, ether and fixed oils

Formula: C₁₆H₁₉N₃O₄S
MW: 349.4 g/mol
Melting Pt: 208 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005175
CAS Number: 69-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fda2d33371f5c8b1c4aad74fd2a0bd555f2f44e&vid=7fda2d33371f5c8b1c4aad74fd2a0bd555f2f44e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR