3-Methylcyclopentadecanone 95, Liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3-Methylcyclopentadecanone, Muscone

0215918680 0215918690
IC15918680EA 238.81 USD
IC15918680 IC15918690
3-Methylcyclopentadecanone 95, Liquid
3-Methylcyclopentadecanone

Insoluble in water. Soluble in ethanol (25 mg/mL) and acetone.

Formula: C₁₆H₃₀O
MW: 238.41
Boiling Pt: 80
Melting Pt: -15
Density: 0.9268
MDL Number: MFCD00211114
CAS Number: 541-91-3
Merck Index: 12,06392

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR