Fuchsin (acid), powder C.I. 42685

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: CI 42685

0215819750 0215819780
IC15819750EA 157.64 USD
IC15819750 IC15819780
Fuchsin (acid), powder C.I. 42685
Fuchsin (acid)

Acid Fuchsin is one of the dyes used in Masson′s Trichome Stain where it is used to distinguish muscle from collagen.


Soluble in water.


Acid Fuchsin is one of the dyes used in Masson′s Trichome Stain where it is used to distinguish muscle from collagen.

Formula: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
MDL Number: MFCD00013286
CAS Number: 3244-88-0

Order Now

Specification Test Results

Identity Test Passes
UV/VIS: l max (water) 543 - 547 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR