N,N'-Octamethylenebis(2,2-dichloroacetamide), off-white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0215805083
0215805091
IC15805083EA
144.85
USD
IC15805083
IC15805091
N,N'-Octamethylenebis(2,2-dichloroacetamide), off-white powder
N,N'-Octamethylenebis(2,2-dichloroacetamide)
N,N'-Octamethylenebis(2,2-dichloroacetamide) is an inhibitor of aldehyde dehydrogenase, ALDH1A2.
Formula:
C₁₂H₂₀Cl₄N₂O₂ Melting Pt: 128.0…128.4 °C (lit.) |
MDL Number:
MFCD00000841 CAS Number: 1477-57-2 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...