Benzocaine (ethyl-4-aminobenzoate), white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215796780 0215796790
IC15796780EA 94.88 USD
IC15796780 IC15796790
Benzocaine (ethyl-4-aminobenzoate), white crystalline powder
Benzocaine (ethyl-4-aminobenzoate)
Formula: C₉H₁₁NO₂
Melting Pt: 88…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR