Diatomaceous earth, powder, acid-washed

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215760791 0215760794 0215760780
IC15760791EA 181.64 USD
IC15760791 IC15760794 76102-758
Diatomaceous earth, powder, acid-washed
Diatomaceous earth

Diatomaceous earth acid-washed is a highly insoluble, inert material and is used as a filtration aid. It is suitable for most routine filtrations.


  • Specific Gravity: 2.3
  • pH: 7.0


Diatomaceous earth acid-washed is used as a filtration aid. It is suitable for most routine filtrations.

Formula: SiO₂
Melting Pt: 1400 °C (lit.)
Density: 13.0 lbs/ft³ (dry)
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 61790-53-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=782afd01e40fa7254a5017cfad511fbd830d219a&vid=54784632119d7f89fef47982942ed2e67be806ce&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=511f96a2bf0516ab99766e55aff53a8c7ea835b6&vid=214843a9a330b06d91eac23eb59f879fb9c9f9a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR