2,2,4,4,6,8,8-Heptamethylnonane, clear, colorless liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0215732225 0215732280
IC15732225EA 66.03 USD
IC15732225 IC15732280
2,2,4,4,6,8,8-Heptamethylnonane, clear, colorless liquid
2,2,4,4,6,8,8-Heptamethylnonane

2,2,4,4,6,8,8-Heptamethylnonane was used as the carrier phase to avoid toxic effects of the pure substances. It is also used as a detergent cleansing agent, emulsifier, and thickening agent in cosmetics.


2,2,4,4,6,8,8-Heptamethylnonane was used as the carrier phase to avoid toxic effects of the pure substances. It is also used as a detergent cleansing agent, emulsifier, and thickening agent in cosmetics.

Formula: C₁₆H₃₄
MDL Number: MFCD00008856
CAS Number: 4390-04-9
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR