2,6-Di-O-methyl-β-cyclodextrine, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, Heptakis(2,6-di-o-methyl)-β-cyclodextrin, 2,6-Di-O-methyl-β-cyclodextrin
Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent.
Heptakis(2,6-Di-o-methyl)-beta-cyclodexrin is used as reactant involved in Physicochemical studies of inclusion complexes, Probing diffusion and single molecule interactions with reconstituted membrane proteins, Interactions with micelles causing micellar rupture. In Studying cellular transport, used as a cell penetration enhancer,Physicochemical and biopharmaceutical improvement of drugs particularly with solubilization and stability, Interactions with cholesterol.
Soluble in water and ethanol.
Heptakis(2,6-Di-o-methyl)-beta-cyclodexrin is used as reactant involved in Physicochemical studies of inclusion complexes, Probing diffusion and single molecule interactions with reconstituted membrane proteins, Interactions with micelles causing micellar rupture. In Studying cellular transport, used as a cell penetration enhancer,Physicochemical and biopharmaceutical improvement of drugs particularly with solubilization and stability, Interactions with cholesterol.
Formula:
C₅₆H₉₈O₃₅ MW: 1331.3563 g/mol |
MDL Number:
MFCD00011616 CAS Number: 51166-71-3 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Reference |
Melting Point | Record Result |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...