Tobramycin, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol

0215692625 0215692680
IC15692625EA 56.02 USD
IC15692625 IC15692680
Tobramycin, powder
Tobramycin

Tobramycin is an aminoglycoside antibiotic derived from Streptomyces tenebrarius and used to treat various types of bacteria infections, particularly Gram-negative infections. It is especially effective against species of Pseudomonas. It works by binding to a site on bacterial 30S and 50S ribosome, preventing formation of the 70S complex. As a result, mRNA cannot be translated into protein and cell death ensues. Tobramycin is preferred over gentamicin for Pseudomonas aeruginosa pneumonia due to better lung penetration.


  • Presentation: White Powder
  • Soluble in water (50 mg/mL).


Tobramycin is an aminoglycoside, broad-spectrum antibiotic produced by Streptomyces tenebrarius. It is effective against gram-negative bacteria, especially the pseudomonas species. It is not effective against Enterococci. It is used to inhibit bacterial protein synthesis. It is used to treat pseudomonas aeruginosa lung infections and is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis, and other soft-tissue infections. It is a potential therapy for sinus infections.


Tobramycin is an aminoglycoside. Its Mode of Action is: it binds to 70S ribosomal subunit; inhibits translocation;and hence elicits miscoding. Spectrum of Activity: Gram negative bacteria.


Tobramycin is an aminoglycoside, broad-spectrum antibiotic produced by Streptomyces tenebrarius


It is effective against gram-negative bacteria, especially the pseudomonas species. It is not effective against Enterococci. It is used to inhibit bacterial protein synthesis. It is used to treat pseudomonas aeruginosa lung infections and is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis, and other soft-tissue infections. It is a potential therapy for sinus infections.


Soluble in water (50 mg/mL)

Formula: C₁₈H₃₇N₅O₉
MW: 467.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077885
CAS Number: 32986-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=835f0481c0881229509611aa22021c01656b3f1e&vid=835f0481c0881229509611aa22021c01656b3f1e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ffc29c974256d3fb3f40aa9e34239cb52118f3e&vid=8ffc29c974256d3fb3f40aa9e34239cb52118f3e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR