Potassium dichromate, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0215633890 0215633891
IC15633890EA 90.27 USD
IC15633890 IC15633891
Potassium dichromate, crystalline powder
Potassium dichromate

Soluble in water (200 mg plus 4 mL).

Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3087
ADR: 5.1 II
Merck Index: 12,07790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR