Ficoll 400, White powder, MP Biomedicals

Supplier: MP Biomedicals
0215632810 0215632825 0215632880
IC15632810EA 114.3 USD
IC15632810 IC15632825 IC15632880
Ficoll 400, White powder, MP Biomedicals
Cell Separation Media
Polysucrose is water-soluble, nontoxic, resistant to intestinal enzymes, spherical and can be produced with a molecular weight distribution that relates to the size of many normal food proteins. It is a nonionic synthetic polymer of sucrose.

  • MW: 400000 g/mol

Polysucrose 400 is used for cell separation and organelle isolation. It has been used in a study to develop a new method for the purification of human motile spermatozoa. It has also been used in a study that investigated the antiproliferative and antitumor activities of D-reverse peptides.

Store at Room Temperature (15 - 30 °C)

Soluble in 100 mg/mL Water.

Polysucrose is water-soluble, nontoxic, resistant to intestinal enzymes, spherical and can be produced with a molecular weight distribution that relates to the size of many normal food proteins. It is a nonionic synthetic polymer of sucrose.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR