Piperacillin sodium salt, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Piperacillin is a semisynthetic, broad-spectrum ureidopenicillin antibiotic. It is derived from ampicillin. Piperacillin inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. Piperacillin may interfere with autolysin inhibitors.
It has been used in pharmacokinetic studies in order to optimize antimicrobial therapy in patients with sepsis. It is used to study piperacillin hypersensitivity reactions and to study multidrug-resistant organisms.
Piperacillin inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. Piperacillin may interfere with autolysin inhibitors.
- Presentation: White Powder
- Soluble in aqueous solution (10 mg/mL-clear, colorless solution), in water (50 mg/mL-clear colorless to faint yellow solution).
- Storage temperature: Store at + 4 °C
Piperacillin sodium salt has been used in pharmacokinetic studies in order to optimize antimicrobial therapy in patients with sepsis. It is used to study piperacillin hypersensitivity reactions and to study multidrug-resistant organisms.
Formula:
C₂₃H₂₇N₅O₇S MW: 539.54 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00056867 CAS Number: 59703-84-3 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...