Paromomycin sulfate ≥97%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate(1:x)

0215605601 0215605605 0215605625
IC15605601EA 99.56 USD
IC15605601 IC15605605 IC15605625
Paromomycin sulfate ≥97%
Paromomycin sulfate

Paromomycin is a broad spectrum aminoglycoside antibiotic produced by Streptomyces rimosus var


  • White powder


Paromomycin is effective against gram-negative bacteria, gram-positive bacteria, some protozoan species, and limited antihelminth. It is used to study bacterial protein synthesis at the level of 16S ribosomal RNA and 30S ribosome assembly. Paromomycin is used to study cytosine-cytosine (CC) mismatch-containing RNA molecules and is used to inhibit Cryptosporidium infection of a human enterocyte cell line.


paromomycinus which has in vitro and in vivo activity similar to neomycin.


Soluble in water (25% w/v - light opalescent solution)

Formula: C₂₃H₄₅N₅O₁₄·H₂SO₄
MW: 615.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079278
CAS Number: 1263-89-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58377e389d8655b613307bde1d7d17c02bbf1f72&vid=58377e389d8655b613307bde1d7d17c02bbf1f72&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR