Pancuronium bromide, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215605310 0215605350 0215605380
IC15605310EA 97.41 USD
IC15605310 IC15605350 IC15605380
Pancuronium bromide, Powder
Pancuronium bromide

Aminosteroidal neuromuscular blocking agent; skeletal muscle relaxant


Pancuronium bromide is used as an adjunct to general anesthesia to facilitate tracheal intubation and to provide skeletal muscle relaxation during surgery or mechanical ventilation.


Very soluble to freely soluble in water (100 mg/mL - clear light yellow solution), very soluble in methylene chloride, freely soluble in alcohol, soluble in chloroform.


Pancuronium bromide is used as an adjunct to general anesthesia to facilitate tracheal intubation and to provide skeletal muscle relaxation during surgery or mechanical ventilation.

Formula: C₃₅H₆₀Br₂N₂O₄
MDL Number: MFCD00079223
CAS Number: 15500-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR