2-Nitro-5-thiocyanatobenzoic acid, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215592983 0215592991
IC15592983EA 139.91 USD
IC15592983 IC15592991
2-Nitro-5-thiocyanatobenzoic acid, crystalline powder
2-Nitro-5-thiocyanatobenzoic acid

2-nitro-5-thiocyanatobenzoic acid (NTCB) is commonly used to cyanylate and cleave proteins at cysteine residues.

Formula: C₈H₄N₂O₄S
MDL Number: MFCD00007139
CAS Number: 30211-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR