p-Nitrophenyl-N-acetyl-α-D-glucosaminide, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215590110 0215590125
IC15590110EA 179.12 USD
IC15590110 IC15590125
p-Nitrophenyl-N-acetyl-α-D-glucosaminide, white powder
p-Nitrophenyl-N-acetyl-α-D-glucosaminide

p-Nitrophenyl-N-acetyl-α-D-glucosaminide is commonly used as a chromogenic substrate for N-acetyl-β-D-glucosaminidase, yielding a yellow solution upon cleavage.


Soluble in water, methanol.

Formula: C₁₄H₁₈N₂O₈
Storage Temperature: Freezer
CAS Number: 10139-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR