3-Nitrophenol, crystals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: m-Nitrophenol

0215589325 0215589380
IC15589325EA 215.94 USD
IC15589325 IC15589380
3-Nitrophenol, crystals
3-Nitrophenol

Soluble in ethanol and water.

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 271
Storage Temperature: Ambient
MDL Number: MFCD00007240
CAS Number: 554-84-7
UN: 1663
ADR: 6.1 III
Merck Index: 13,06652
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cad0ee990a953f34599fcb1922478f60df77a06&vid=7cad0ee990a953f34599fcb1922478f60df77a06&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR