Nitrobenzene, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215585190
0215585191
IC15585190EA
70.23
USD
IC15585190
IC15585191
Nitrobenzene, liquid
Nitrobenzene
Soluble in alcohol, benzene, oils, methanol, acetone and diethyl ether; slightly soluble in cold water.
Formula:
C₆H₅NO₂ MW: 123.11 g/mol |
MDL Number:
MFCD00007043 CAS Number: 98-95-3 UN: 1662 ADR: 6.1,II |
Specification Test Results
Form | Yellow Liquid |
Storage | Store at Room Temperature (15-30 °C). |
Refractive Index | n20/D 1.551(Lit.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...