2-Methyl-5-nitroimidazole-1-ethanol, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Metronidazole

0215571005 0215571025
IC15571005EA 104.69 USD
IC15571005 IC15571025
2-Methyl-5-nitroimidazole-1-ethanol, white powder
2-Methyl-5-nitroimidazole-1-ethanol

Metronidazole ia an antiprotozoal (Trichomonas), a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria.


Metronidazole suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. Further studies show that Metronidazole has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such as peptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration. Furthermore, the gene hefA codes for the TolC efflux pump in Helicobacter pylori which is resistant to Metronidazole.


Soluble in water (10 mg/ml) or dilute acids (50 mg/ml 0,1 M acetic acid - clear to slightly hazy, light yellowish-green solution); slightly soluble in ethanol (5 mg/ml).


Metronidazole ia an antiprotozoal (Trichomonas), a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria.

Formula: C₆H₉N₃O₃
MW: 171.16 g/mol
Melting Pt: 316…320° F
Storage Temperature: Refrigerator
MDL Number: MFCD00009750
CAS Number: 443-48-1

Order Now

Specification Test Results

Synonyms 2-Methyl-5-nitroimidazole-1-ethanol; 1-(2-Hydroxyethyl)-2-methyl-5-nitroimidazole; 1-(b-Ethylol)-2-methyl-5-nitro-3-azapyrrole
Form White Powder
Storage Store at +4 °C.
Loss on Drying ≤2%
pH 5 - 7(1% Aqueous solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR