O-Methylhydroxylamine hydrochloride, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

0215540501 0215540525
IC15540501EA 35 USD
IC15540501 IC15540525
O-Methylhydroxylamine hydrochloride, crystalline powder
O-Methylhydroxylamine hydrochloride

Soluble in water, alcohol.

Formula: CH₆ClNO
MW: 83.515 g/mol
MDL Number: MFCD00012951
CAS Number: 593-56-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR