Malt extract, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215531580
0215531590
IC15531580EA
110.46
USD
IC15531580
IC15531590
Malt extract, Powder
Malt extract
Soluble in water (50 mg/mL ).
Malt extract is a source of carbohydrates.
MDL Number:
MFCD00131570 CAS Number: 8002-48-0 |
Specification Test Results
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). Store dessicated. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...