Lithium iodide, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215525910 0215525950 0215525983
IC15525910EA 113.49 USD
IC15525910 IC15525950 IC15525983
Lithium iodide, Powder
Lithium iodide

Lithium iodide controls regioselectivity in palladium-catalyzed allylic alkylation reactions.

Formula: ILi
MW: 133.85 g/mol
MDL Number: MFCD00011092
CAS Number: 10377-51-2

Order Now

Specification Test Results

Form Off-white Powder
Storage Store at room temperature (15-30 °C), desiccate, protect from light.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR