(R)-(+)-Limonene, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: (R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene

0215523480 0215523490
IC15523480EA 81.65 USD
IC15523480 IC15523490
(R)-(+)-Limonene, liquid
(R)-(+)-Limonene

D-Limonene is a chiral intermediate for natural product synthesis. It is used as a renewably based solvent in cleaning products.


D-Limonene is a monoterpene found to prevent mammary cancer by inducing hepatic glutathione-S- transferase and uridine diphosphoglucuronosyl transferase. D-Limonene is an isoprenylation inhibitor of small G-proteins.


Limonene is a cyclie terpene from Chinese medicinal herb essential oils used in the synthesis of carvone. Limonene may be used as a shrinking agent to dissolve polystyrene. Limonene may be used in various insecticidal and insect repellant applications.


Miscible with alcohol, with most fixed oils, mineral oil; very slightly soluble in glycerin; practically insoluble in water and in propyiene glycol.


D-Limonene is a monoterpene found to prevent mammary cancer by inducing hepatic glutathione-S- transferase and uridine diphosphoglucuronosyl transferase. D-Limonene is an isoprenylation inhibitor of small G-proteins.

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Density: 0.84 g/ml
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
UN: 2052
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b58d6ffbe1286e81dff1578b60b004f1d0bbd2a&vid=8b58d6ffbe1286e81dff1578b60b004f1d0bbd2a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR