Leuco crystal violet, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 4,4',4''-Methanetriyltris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane, 4,4',4''-Methylidynetris(N,N-dimethylaniline)

0215519505 0215519525
IC15519505EA 55.92 USD
IC15519505 IC15519525
Leuco crystal violet, powder
Leuco crystal violet

Soluble in benzene (0.5/20-clear).


  • λmax 260 nm(lit.)


Precaución: Research Use Only (RUO).

Formula: C₂₅H₃₁N₃
MW: 373.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008314
CAS Number: 603-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR