Lead tetraacetate ≥95%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215518225 0215518280
IC15518225EA 59.49 USD
IC15518225 IC15518280
Lead tetraacetate ≥95%, powder
Lead tetraacetate

Lead tetraacetate Smoothly induces the addition of difluorodiiodomethane to alkenes and alkynes.

Formula: (H₃CCOO)₄Pb
MDL Number: MFCD00008693
CAS Number: 546-67-8
UN: 1616
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR