Kojic acid, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215515701 0215515705 0215515710
IC15515701EA 58.34 USD
IC15515701 IC15515705 IC15515710
Kojic acid, white crystalline powder
Kojic acid

Kojic acid is used as an antibiotic reagent. It is tyrosinase inhibitor. Kojic acid is a chelation agent.


Soluble in water, ethanol, acetone; Slightly soluble in ethyl acetate, chloroform, pyridine.


Kojic acid is used as an antibiotic reagent.

Formula: C₆H₆O₄
Melting Pt: 148…157 °C
MDL Number: MFCD00006580
CAS Number: 501-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR