Indocyanine green ~90% (dye content)

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen

0215502025 0215502050 0215502080 0215502005
IC15502025EA 112.77 USD
IC15502025 IC15502050 IC15502080 75875-556
Indocyanine green ~90% (dye content)
Indocyanine green

Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.


  • Extinction Coefficient (E1%): 10.7-11.6 (393-394 nm)(Lit.)


It is used for determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. ICG binds tightly to plasma proteins and becomes confined to the vascular system.
Indocyanine Green is a negatively charged polymethine dye that forms noncovalent fluorescent complexes with proteins. This "pseudofluorogenic" property has been utilized for protein determination by capillary electrophoresis with diode laser-induced fluorescence detection. ICG is a fluorescent dye which is used in medicine as an indicator substance (e.g. for photometric hepatic function diagnostics and fluorescence angiography) in cardiac, circulatory, hepatic and ophthalmic conditions. It has been used to resolve several labeled proteins including: human serum albumin, ribonuclease A, transferrin, and cytochrome c.


Sparingly soluble in acetone/glycil ethers and ehtanol; water (10 mg/1mL - transparent solution.)

Formula: C₄₃H₄₇N₂NaO₆S₂
MW: 774.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013078
CAS Number: 3599-32-4
EINECS: 222-751-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c12bcfb8dd0da6ff209f36268c45d49baa1db96&vid=8c12bcfb8dd0da6ff209f36268c45d49baa1db96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR