Imidazolidinyl urea, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Imidurea, 1,1'-Methylenebis(3-(3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)urea), 1,1'-Methylenebis[3-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]urea], N,N'-Methylenebis[N'-(3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl)urea]

0215500980 0215500983 0215500925
IC15500980EA 119.13 USD
IC15500980 IC15500983 IC0215500925
Imidazolidinyl urea, white powder
Imidazolidinyl urea

Imidazolidinyl urea is an antimicrobial preservative.


It is topical formulas for use as antioxidants It is used in various studies such as Biological studies investigating the antibacterial synergistic activity of essential oils and surfactants against P. aeruginosa and S. aureus, Pharmacological studies investigating nutrient-sensitized screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis, Therapeutic studies involving nanostructured vegetable-based carriers for topical delivery of active molecules, In comparison studies of MEKC and microemulsion electrokinetic chromatography for analytical studies of preservatives, Investigations of in vitro induction of apoptosis versus necrosis.


Solubility : Soluble in water(50 mg/mL).


Imidazolidinyl urea is an antimicrobial preservative.


Precaución: Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Formula: C₁₁H₁₆N₈O₈
MW: 388.30 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00221482
CAS Number: 39236-46-9

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR