Chromeazurol S (Mordant blue 29), dark brown powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 2,6-Dichlorodimethyl-sulphoxy-fuchsonedicarboxylic acid sodium salt, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid,trisodium salt, Mordant Blue 29, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid, trisodium salt, Chrome Azurol S

0215498225 0215498280 0215498210
IC15498225EA 106.47 USD
IC15498225 IC15498280 76102-610
Chromeazurol S (Mordant blue 29), dark brown powder
Chromeazurol S (Mordant blue 29)

Chrome Azurol S is used as a reagent for determination of fluoride.


  • Presentation: Dark Brown Powder
  • UV/Visible Absorbance: λmax 458 nm (Lit.)
  • Soluble in water, in most organic solvents.
  • Storage temperature: Store at Room Temperature (15-30 °C)


Chrome Azurol S is used as a reagent for determination of fluoride.

Formula: C₂₃H₁₆Cl₂O₉S
Storage Temperature: Ambient
MDL Number: MFCD00001615
CAS Number: 1667-99-8

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR