β-Caryophyllene, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215494405 0215494410 0215494425
IC15494405EA 194.36 USD
IC15494405 IC15494410 IC15494425
β-Caryophyllene, liquid
β-Caryophyllene

(-)-trans-Caryophyllene is a natural bicyclic sesquiterpene that is a constituent of many essential oils.

Formula: C₁₅H₂₄
MDL Number: MFCD00075925
CAS Number: 87-44-5

Order Now

Specification Test Results

Synonyms β-Caryophyllene; trans-(1R,9S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene
Form Clear, Slight Yellow Liquid
Storage Store at Room Temperature (15-30 °C).
Optical Rotation [α]20/D -10±1 °, neat (Lit.)
Refractive Index n20/D 1.499 (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR