Bronopol, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0215488725 0215488780
IC15488725EA 42.11 USD
IC15488725 IC15488780
Bronopol, white crystalline powder
Bronopol

Soluble in a 1% aqueous solution (clear, colorless solution).


Soluble in a 1% aqueous solution (clear, colorless solution).


Precaución: Research Use Only (RUO).

Formula: HOCH₂BrC(NO₂)CH₂OH
Melting Pt: 125…135 °C
MDL Number: MFCD00007390
CAS Number: 52-51-7
UN: 3241
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20396db6a2c90777734fd529024a0aa4b4bdc37b&vid=20396db6a2c90777734fd529024a0aa4b4bdc37b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR