D-(+)-Trehalose dihydrate ≥99%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
Trehalose is a non-reducing sugar formed from two glucose units joined by a 1-1 alpha bond. It is thought to provide plants and animals with the ability to withstanding periods of dehydration.
Use as a cryoprotectant in a variety of cell freezing media.
Precaución: Research Use Only (RUO).
Formula:
C₁₂H₂₂O₁₁·2H₂O MW: 378.33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00071594 CAS Number: 6138-23-4 |
Specification Test Results
Identity Test | Passes |
Purity | #99% |
FTIR | Conforms to Standard |
Loss on Drying | 8.5 - 10.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...