Methyl-ɑ-D-mannopyranoside ∼99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

0215393710 0215393725 0215393780
IC15393710EA 42.83 USD
IC15393710 IC15393725 IC15393780
Methyl-ɑ-D-mannopyranoside ∼99%, white crystalline powder
Methyl-alpha-D-mannopyranoside

Used to reduce Con A binding to lymphocyte surface in culture.


Methyl α-D-mannopyranoside has been used to synthesize a series of tri- and tetrahydroxylated seven-membered iminosugars in a study that worked towards a stable noeuromycin analog with a D-manno configuration. It has also been used in a study to investigate the primary mannose binding site of pradimicin A.


Soluble in water.


Used to reduce Con A binding to lymphocyte surface in culture. Methyl α-D-mannopyranoside has been used to synthesize a series of tri- and tetrahydroxylated seven-membered iminosugars in a study that worked towards a stable noeuromycin analog with a D-manno configuration. It has also been used in a study to investigate the primary mannose binding site of pradimicin A.


Precaución: Research Use Only (RUO).

Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063262
CAS Number: 617-04-9

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 187 - 197 °C
Loss on Drying ≤0.5%
Optical Rotation +76 to +85 °C (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR