(6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
(6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is necessary in vivo for the conversion of phenylalanine to tyrosine and for the production of the hormone epinephrine and of the monoamine neurotransmitters, serotonin, dopamine, and norepinephrine. It is also involved in apoptosis and other cellular events mediated by nitric oxide production and catalyzes the cleavage of alkylglycerol ethers.
Natural cofactor for phenylalanine hydroxylase, tyrosine hydroxylase, tryptophan hydroxylase, and nitric oxide synthetase.
Stability: (6R)-BH4 . 2HCl is hygroscopic. It reacts with oxygen especially in neutral and basic solutions, but dry, in ampules and at -20 °C or colder it can be stored for several years. Oxidized tetrahydrobiopterin solutions become yellow but are relatively stable.
Soluble in water (2.3 g completely soluble in 100 mL). A 1 mM solution in water results in a pH of 3.0 and 1 M solution in water results in a pH of 0.45.
(6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is necessary in vivo for the conversion of phenylalanine to tyrosine and for the production of the hormone epinephrine and of the monoamine neurotransmitters, serotonin, dopamine, and norepinephrine. It is also involved in apoptosis and other cellular events mediated by nitric oxide production and catalyzes the cleavage of alkylglycerol ethers.
Formula:
C₉H₁₅N₅O₃·2HCl |
MDL Number:
MFCD00891665 CAS Number: 17528-72-2 |
Specification Test Results
Synonyms | (6R)-BH4-Amino-6-(1,2-dihydroxy-propyl)-5,6,7,8-tetrahydro-4(1H)-pteridinone dihydrochloride; BH4; (6R)-BH4; (6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride; Sapropterin; Dapropterin |
Form | White Powder |
Storage | Store at -20 °C. Store Desiccated. |
Extinction Coefficient | EM(216 nm)=15,400 (0.1 N HCl); (264 nm) = 13,750 (0.1 N HCl)(Lit.) |
Optical Rotation | [α]D = -6.6° to -7.6° (C = 1 in 0.1 N HCl)(Lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...