Hydroxypropyl-β-cyclodextrin, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
HBC, 2-Hydroxypropylether-b-cyclodextrin
2-Hydroxypropyl-beta-cyclodextrin are non-toxic solubilisers. The solubility of drugs increases linearly with the concentration of 2-Hydroxypropyl-b-cyclodextrin in aqueous buffer. The formation of drug/cyclodextrin complexes is a rapidly reversible reaction and complexes exist both in solution and crystalline states.
Solutions of many such complexes may be lyophilised to produce freely soluble powders which may be compressed into tablets. Bio-effects are only slightly affected by cyclodextrin complexation. Cells in serum supplemented medium can be grown in up to 1 to 2%. In serum-free medium, concentrations of 0,5 to 1% are acceptable. Hydroxypropyl-b-cyclodextrin has been found to be non-toxic in mice and rabbits. 2-hydroxypropyl-g-cyclodextrin with its 8 glucose units, has a slightly larger cavity and can accomodate larger substrates than the beta form. The use of cyclodextrins in receptor binding assays in not recommended.
Soluble in water (50 g/100 mL). Solutions may be obtained by stirring 30 minutes at room temperature. Alternatively, sonication with cooling may be employed. Solutions may be stored for several weeks at room temperature. Good solubility in methanol, ehtanol, pyridine, dimethyl sulfoxide, dimethyl formamide.
2-Hydroxypropyl-beta-Cyclodextrin are non-toxic solubilizers. The solubility of drugs increases linearly with the concentration of 2-Hydroxypropyl-b-cyclodextrin in aqueous buffer. The formation of drug/cyclodextrin complexes is a rapidly reversible reaction and complexes exist both in solution and crystalline states. Solutions of many such complexes may be lyophilized to produce freely soluble powders which may be compressed into tablets. Bio-effects are only slightly affected by cyclodextrin complexation. Cells in serum supplemented medium can be grown in up to 1-2%. In serum-free medium, concentrations of 0.5-1% are acceptable. Hydroxypropyl-b-cyclodextrin has been found to be non-toxic in mice and rabbits. 2-hydroxypropyl-g-cyclodextrin with its 8 glucose units, has a slightly larger cavity and can accomodate larger substrates than the beta form. The use of cyclodextrins in receptor binding assays in not recommended.
Formula:
C₅₄H₁₀₂O₃₉ MW: 1375.371 g/mol |
MDL Number:
MFCD16621721 CAS Number: 94035-02-6 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...