Sodium hydroxide ≥97%, pellets
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Caustic soda
1g dissolves in 0.9 mL water; 0.3 mL boiling water, 7.2 mL absolute alcohol, 4.2 mL methanol; insoluble in glycerol. The dissolution of sodium hydroxide in water is a highly exothermic reaction. Sodium hydroxide stock solutions should be stored in plastic containers. Glass containers should be avoided in the preparation and storage of sodium hydroxide solutions because it will etch the glass (typically anything over 2 N will etch glass). Keep solutions protected from air as much as possible since contact with air will form sodium carbonate.
Poison/Deleterious Substance Code: 2-54
Formula:
NaOH MW: 40 g/mol |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...