Thioacetamide ≥99.0%, crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Ethanethioamide

0215258725 0215258780
IC15258725EA 123.77 USD
IC15258725 IC15258780
Thioacetamide ≥99.0%, crystalline powder ACS
Thioacetamide

Thioacetamide is a hepatotoxin and a carcinogen. Thioacetamide is also an additive in enantioselective reduction of β-keto esters with immobilized bakers yeast.


Thioacetamide is a source of hydrogen sulphide in qualitative analysis. It is a potent hepatotoxicant.


Thioacetamide is a source of hydrogen sulfide in qualitative analysis. It is a potent hepatotoxicant.

Formula: CH₃CSNH₂
MW: 75.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008070
CAS Number: 62-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay >99,0%
Melting Point 111 - 114°C
Residue on Ignition <0,05%
Clarity of a 2% Solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR