D-(+)-Sucrose, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

0215258490 0215258491
IC15258490EA 86.69 USD
IC15258490 IC15258491
D-(+)-Sucrose, white crystalline powder ACS
D-(+)-Sucrose

Simple carbohydrate. Sucrose is hydrolyzed to glucose and fructose by dilute acids and by invertase, a yeast enzyme. Upon hydrolysis the optical rotation falls and is negative when the hydrolysis is complete. The mixture of glucose and fructose is known as "Invert sugar".


Sucrose can be used to prepare density gradients for cell organelle separation. Sucrose is a main component of culture media. It can also be used in various enzymatic assays.


Soluble in water (1 g/0.5 mL at 25°C; 1 g/0.2 mL boiling water), ethanol (1 g/170 mL) or methanol (1 g/100 mL); moderately soluble in glycerol or pyridine.


Sucrose can be used to prepare density gradients for cell organelle seperattion. Sucrose is a main component of culture media. It can also be used in various enzymatic assays

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 185…187 °C (lit.)
Density: 1.587 (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Rotation (c=26, water) +66.3° to +66.8°
Insoluble Matter ≤0.03%
Loss on Drying (105°C) ≤0.03%
Residue on Ignition ≤0.01%
Titratable Acid ≤0.0008 meq/g
Chloride (Cl) ≤0.005%
Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤5 ppm
Invert Sugar ≤0.05%
Sulfate and Sulfite (as SO4) ≤0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR