Sodium bisulfite, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Sodium hydrogen sulphite
0215257380
0215257390
IC15257380EA
43.66
USD
IC15257380
IC15257390
Sodium bisulfite, powder ACS
Sodium bisulphite
Chloride: ≤0.02%, Iron: ≤0.002%
Formula:
HNaO₃S MW: 104.06 g/mol |
MDL Number:
MFCD00167602 CAS Number: 7631-90-5 UN: 2693 ADR: 8,III |
Specification Test Results
Synonyms | Sodium hydrogensulfite |
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). |
Assay | ≥58.5% SO2 |
Heavy Metals | ≤0.001% |
Insoluble Matter | ≤0.005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...