L(+)-Potassium sodium tartrate tetrahydrate ≥99.0%, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215256380 0215256390 0215256392
IC15256380EA 58.94 USD
IC15256380 IC15256390 76102-746
L(+)-Potassium sodium tartrate tetrahydrate ≥99.0%, white crystalline powder ACS
L(+)-Potassium sodium tartrate tetrahydrate

Potassium sodium tartrate tetrahydrate has been used in organic synthesis to break up emulsions in aqueous workups.


  • Refractive Index: 22 ° (C=10, H₂O) (lit.)


The compound is an ingredient of Fehling’s solution and Biuret reagent and is used in detection of aldose and ketose functional group and measuring of protein concentrations. Potassium sodium tartrate tetrahydrate is a component in the technique to produce micrometer sized aerosol particles. The compound has been used as a precipitant in the crystallization and preliminary X-ray analysis of strictosidine synthase.


Chloride- Not more than 0.001%, Sulfate- Not more than 0.005%, Ammonium - Not more than 0.002% ,Iron- Not more than 0.001%,Phosphate Not more than 0.002% ,Calcium- Not more than 0.005%


Potassium sodium tartrate tetrahydrate has been used in organic synthesis to break up emulsions in aqueous workups. The compound is an ingredient of Fehling′s solution and Biuret reagent and is used in detection of aldose and ketose functional group and measuring of protein concentrations. Potassium sodium tartrate tetrahydrate is a component in the technique to produce micrometer sized aerosol particles. The compound has been used as a precipitant in the crystallization and preliminary X-ray analysis of strictosidine synthase.


Precaución: Research Use Only (RUO).

Formula: KOOC(CHOH)₂COONa·4H₂O
Boiling Pt: 399.3 °C (760 mmHg)) (lit.)
Melting Pt: 70 °C (lit.)
Density: 1.05 g/mL (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656ffd1a6d86fe3aa99b4a7163c275bc1b186a5a&vid=656ffd1a6d86fe3aa99b4a7163c275bc1b186a5a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (non-aq titr) ≥99.0%
pH (5% aq soln) 6.0-8.5
Insoluble Matter ≤0.005%
Chloride ≤0.001%
Phosphate ≤0.002%
Sulfate ≤0.005%
Ammonium ≤0.002%
Calcium ≤0.005%
Heavy Metals ≤5ppm
Iron ≤0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR