Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, yellow crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate

0215256080 0215256090
IC15256080EA 78.93 USD
IC15256080 IC15256090
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, yellow crystalline powder ACS
Potassium hexacyanoferrate(II) trihydrate

Potassium ferrocyanide trihydrate is a salt of the Fe(CN)₆4- ferrocyanide ion. Ferrocyanide is a reducing iron species, being oxidized from ferrous Fe(II) to ferric Fe(III).


Potassium ferrocyanide is used to determine the concentration of potassium permanganate, a compound often used in titrations based on redox reactions. Potassium ferrocyanide is used in a mixture with potassium ferricyanide and phosphate buffered solution (PBS) to provide a buffer for X-Gal, which is used to cleave Beta-galactosidase, giving a bright blue visualization where an antibody (or other molecule), conjugated to Beta-gal, has bonded to its target.


The ferrocyanide species is membrane impermeable and can be used to assess redox activity of membrane-bound structures and mechanisms of electron transport across membranes. This approach was employed to demonstrate that Potassium ferrocyanide performs reducing activation of membrane-bound dopamine β-monooxygenase from the exterior face of adrenal vesicles, providing elucidation of native reduction-activation mechanisms for dopamine β-monooxygenase regulation.


Chloride ≤0.01%


Potassium ferrocyanide is used to determine the concentration of potassium permanganate, a compound often used in titrations based on redox reactions. Potassium ferrocyanide is used in a mixture with potassium ferricyanide and phosphate buffered solution (PBS) to provide a buffer for X-Gal, which is used to cleave Beta-galactosidase, giving a bright blue visualization where an antibody (or other molecule), conjugated to Beta-gal, has bonded to its target.

Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 70 (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
Merck Index: 12,07793

Order Now

Specification Test Results

Assay 98.5 - 102.0%
Insoluble Matter ≤0.005%
Chloride ≤0.01%
Sulfate Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR