Dodecamolybdophosphoric acid hydrate, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate
0215255625
0215255680
IC15255625EA
134.68
USD
IC15255625
IC15255680
Dodecamolybdophosphoric acid hydrate, powder ACS
Dodecamolybdophosphoric acid hydrate
Phosphomolybdic acid hydrate is employed in preparation of a hybrid organic-inorganic, electroactive material with potential application as a cation-insertion electrode in batteries.
Very soluble in alcohol, ether; slightly soluble in less than 0,4 part water.
Precaución: Research Use Only (RUO).
Formula:
H₃[P(Mo₃O₁₀)₄]·nH₂O MW: 1843.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149913 CAS Number: 51429-74-4 UN: 3260 ADR: 8,III |
Specification Test Results
Identity Test | Passes |
Insoluble Matter | ≤0.01% |
Chloride (Cl) | <0,02% |
Sulphate (SO4) | <0,025% |
Ammonium (NH4) | <0,01% |
Calcium (Ca) | <0,02% |
Heavy Metals (as Pb) | <0,005% |
Iron (Fe) | <0,005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...