Nitrilotriacetic acid ≥98%, white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: NTA, 2,2',2''-Nitrilotriacetic acid

0215255180 0215255190 0215255191
IC15255180EA 308.35 USD
IC15255180 IC15255190 IC15255191
Nitrilotriacetic acid ≥98%, white powder ACS
Nitrilotriacetic acid

Nitrilotriacetic acid is an excellent chelating agent which sequester metal ions to form water-soluble complexes; It has many industrial applications. Because of its ability to chelate calcium and magnesium ions.


Soluble in 1M Sodium Hydroxide (100 mg/ml).


Nitrilotriacetic acid is an excellent chelating agent which sequester metal ions to form water-soluble complexes; It has many industrial applications. Because of its ability to chelate calcium and magnesium ions.

Formula: C₆H₉NO₆
MW: 191.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004287
CAS Number: 139-13-9
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Assay ≥98%
Clarity of solution Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR