Lithium chloride ≥99%, white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215253880 0215253890
IC15253880EA 116.26 USD
IC15253880 IC15253890
Lithium chloride ≥99%, white powder ACS
Lithium chloride

Lithium chloride is used as an electrolyte for low temperature dry battery cells and as an oxidation catalyst.


It is a solubilizer for polyamides and cellulose when used with amide solvents, and is a chlorinating agent for steroid substrates. Lithium chloride has also been utilized in: Large scale plasmid DNA isolation without ultracentrifugation, protein extraction and protein crystallization, crystallization of other biological structures including vitamin B12-RNA aptamer and the L-A virus particle, Inhibits the expression and secretion of insulin-like growth factor-binding protein-1 in H4-II-E cells, used in the synthesis of beta-substituted alpha-amino acid derivatives.


Nitrate (NO3) ≤0.001%; Sulfate (SO4) ≤0.01%; Barium (Ba) ≤0.003%; Iron (Fe) 0.001%; Calcium (Ca) ≤0.01%; Potassium (K) ≤0.01%; Sodium (Na) ≤0.20%

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614 (lit.)
MDL Number: MFCD00011078
CAS Number: 7447-41-8
Merck Index: 13,05550

Order Now

Specification Test Results

Assay ≥99%
Insoluble Matter ≤0.01%
Titrable Base ≤0.008 mEq/g
Loss on Drying ≤1.0%
Nitrate (NO₃) ≤0.001%
Sulfate (SO₄) ≤0.01%
Barium (Ba) ≤0.003%
Heavy Metals ≤0.002%
Iron (Fe) <0.001%
Calcium (Ca) ≤0.01%
Potassium (K) ≤0.01%
Sodium (Na) ≤0.20%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR