Gallic acid monohydrate ≥98.0%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate

0215252580 0215252590
IC15252580EA 173.81 USD
IC15252580 IC15252590
Gallic acid monohydrate ≥98.0%, powder ACS
Gallic acid monohydrate

Sulfate (SO4): ≤0.02%

Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
MDL Number: MFCD00149098
CAS Number: 5995-86-8

Order Now

Specification Test Results

Synonyms 3,4,5-Trihydroxybenzoic acid
Form White Powder
Storage Store at Room Temperature (15-30 °C). Store dessicated.
Assay ≥98.0%
Insoluble Matter ≤0.01%
Residue on Ignition ≤0.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR