Dimethyl glyoxime, white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime

0215251825 0215251880
IC15251825EA 52.9 USD
IC15251825 IC15251880
Dimethyl glyoxime, white powder ACS
Dimethyl glyoxime

Soluble in alcohol, ether, pyridine, acetone; practically insoluble in water.


Insoluble In Alcohol: ≤0.05%

Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Melting Pt: ~240 °C
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
UN: 2926
ADR: 4.1,III

Order Now

Specification Test Results

Identity Test Passes
Melting Point ~240°C
Suitability for Ni determination Passes
Insoluble In Alcohol ≤0.05%
Residue on Ignition ≤0.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR