4-(Dimethylamino)benzaldehyde, crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

0215251750 0215251783 0215251710 0215251780
IC15251750EA 90.1 USD
IC15251750 IC15251783 76204-110 76204-112
4-(Dimethylamino)benzaldehyde, crystalline powder ACS
4-(Dimethylamino)benzaldehyde

Soluble in alcohol (40 mg/mL-ethanol), HCl (50 mg/mL-10% HCl), benzene, chloroform, ether; slightly soluble in water.


  • UV/Visible Absorbance : λ min 369 nm; λ max 385 nm(Lit.)
  • Presentation : Off-white Crystalline Powder
  • Color (APHA) : ≤60 of Alcohol Solution
  • Residue on ignition : ≤0.1%
  • Solubility : Soluble in alcohol (40 mg/mL-ethanol), HCl (50 mg/mL-10% HCl), benzene, chloroform, ether; slightly soluble in water.


Storage: protect from light, under nitrogen.

Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176...177 °C (17 mmHg) (lit.)
Melting Pt: 73...75 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Melting Point 73 - 75°C
Solubility in Alcohol Passes
Color (APHA) of Alcohol
Solution
<60
Solubility in Hydrochloric
Acid
Passes
Color of Hydrochloric Acid
Solution
Passes
Residue after Ignition <0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR