Bromophenol blue sodium salt, dark green powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
Soluble in water (30 mg/ml - clear, blueish violet solution) or ethanol (7 mg/ml).
Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
Precaución: Research Use Only (RUO).
Formula:
C₁₉H₉Br₄NaO₅S MW: 691.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013793 CAS Number: 34725-61-6 |
Specification Test Results
Clarity of Solution | Passes |
Visual Transition Interval | pH 3.0 (yellow) to pH 4.6 (blue) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...