Bromocresol green sodium salt, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

0215250305 0215250325
IC15250305EA 65 USD
IC15250305 IC15250325
Bromocresol green sodium salt, powder ACS
Bromocresol green sodium salt

Soluble in water (40 mg/mL) or ethanol (60 mg/mL).


Visual Transition Interval Yellow (pH 3.8) to Blue (pH 5.4).


Bromocresol Green is useful in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualisation of the compounds with functional groups whose pKa is below 5.0.

Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
MDL Number: MFCD00148898
CAS Number: 62625-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR